Product Details

SNP ID
rs17886456
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:55710155 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TACCCAGAAATAACTGCTCTGAATG[C/T]TTGCTACTGTGGAGATGACATGTTT
Phenotype
MIM: 609663
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NLRP9 PubMed Links

Gene Details

Gene
NLRP9
Gene Name
NLR family pyrin domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176820.3 Intron NP_789790.2
XM_011526894.2 Intron XP_011525196.1
XM_011526895.1 Intron XP_011525197.1

View Full Product Details