Product Details
- SNP ID
-
rs7508201
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:8494270 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTCCAGCAGGGCTTCTGGGAGTGG[G/T]TGGAGGCAGGCTGTCACTAGCCCAG
- Phenotype
-
MIM: 160994
MIM: 606466
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HNRNPM
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116059063] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HNRNPM
- Gene Name
- heterogeneous nuclear ribonucleoprotein M
There are no transcripts associated with this gene.
- Gene
- PRAM1
- Gene Name
- PML-RARA regulated adaptor molecule 1
View Full Product Details