Product Details

SNP ID
rs1423004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:148067773 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTAGGGATTATTCTTATTGATG[A/C]AACTCTTTAATTTTTCTTATAAGTA
Phenotype
MIM: 605010
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPINK5 PubMed Links

Gene Details

Gene
SPINK5
Gene Name
serine peptidase inhibitor, Kazal type 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127698.1 Intron NP_001121170.1
NM_001127699.1 Intron NP_001121171.1
NM_006846.3 Intron NP_006837.2
XM_011537551.2 Intron XP_011535853.1

View Full Product Details