Product Details

SNP ID
rs1423002
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:148069943 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAAATGTAGGCAGACATTTTATAAT[A/G]TTCAATGCTCAAACACCTTGCCTCA
Phenotype
MIM: 605010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPINK5 PubMed Links

Gene Details

Gene
SPINK5
Gene Name
serine peptidase inhibitor, Kazal type 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127698.1 Intron NP_001121170.1
NM_001127699.1 Intron NP_001121171.1
NM_006846.3 Intron NP_006837.2
XM_011537551.2 Intron XP_011535853.1

View Full Product Details