Product Details

SNP ID
rs7327284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102604667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTCAGGCTTTTGGCTTAAAACA[A/G]TTCAGTTCTCTTTTTTAAGTGAATG
Phenotype
MIM: 190470
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TPP2 PubMed Links

Gene Details

Gene
TPP2
Gene Name
tripeptidyl peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005254070.1 Intron XP_005254127.1
XM_005254071.1 Intron XP_005254128.1
XM_005254072.1 Intron XP_005254129.1

View Full Product Details