Product Details

SNP ID
rs6671171
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:156901526 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACAAGTTTTGTCCATGCCACCAC[A/G]CCATAGGCTCTGTGAACAGATCCAG
Phenotype
MIM: 610278
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PEAR1 PubMed Links

Gene Details

Gene
PEAR1
Gene Name
platelet endothelial aggregation receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080471.1 Intron NP_001073940.1
XM_005245141.3 Intron XP_005245198.1
XM_011509510.2 Intron XP_011507812.1
XM_011509511.2 Intron XP_011507813.1
XM_011509512.2 Intron XP_011507814.1
XM_017001234.1 Intron XP_016856723.1
XM_017001235.1 Intron XP_016856724.1
XM_017001236.1 Intron XP_016856725.1
XM_017001237.1 Intron XP_016856726.1
XM_017001238.1 Intron XP_016856727.1
XM_017001239.1 Intron XP_016856728.1
XM_017001240.1 Intron XP_016856729.1
XM_017001241.1 Intron XP_016856730.1
XM_017001242.1 Intron XP_016856731.1

View Full Product Details