Product Details
- SNP ID
-
hCV2012036
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:69410866 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAGCTGTAGTTGACACTTTGACT[A/T]CTAAAATGGGCATGGCACAAGGAAA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SLC39A9
PubMed Links
Gene Details
- Gene
- SLC39A9
- Gene Name
- solute carrier family 39 member 9
There are no transcripts associated with this gene.
View Full Product Details