Product Details

SNP ID
rs1410057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110931389 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGCATGTTAATTGTGGGTGTTTT[C/G]TCATTCTTTGGACCAATCTTTTGGT
Phenotype
MIM: 602775
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RPL13AP6 PubMed Links

Gene Details

Gene
RPL13AP6
Gene Name
ribosomal protein L13a pseudogene 6
There are no transcripts associated with this gene.

Gene
SHOC2
Gene Name
SHOC2, leucine rich repeat scaffold protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001269039.2 Intron NP_001255968.1
NM_001324336.1 Intron NP_001311265.1
NM_001324337.1 Intron NP_001311266.1
NM_007373.3 Intron NP_031399.2
XM_017016702.1 Intron XP_016872191.1
XM_017016703.1 Intron XP_016872192.1
XM_017016704.1 Intron XP_016872193.1

View Full Product Details