Product Details

SNP ID
rs10751532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125786138 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTCCCACATTGACTCTGGGAATC[C/T]GGCACCTAGAACATCTACGAACTCA
Phenotype
MIM: 606938
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UROS PubMed Links

Gene Details

Gene
UROS
Gene Name
uroporphyrinogen III synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000375.2 Intron NP_000366.1
NM_001324036.1 Intron NP_001310965.1
NM_001324037.1 Intron NP_001310966.1
NM_001324038.1 Intron NP_001310967.1
NM_001324039.1 Intron NP_001310968.1
XM_005270140.4 Intron XP_005270197.1
XM_011540127.1 Intron XP_011538429.1
XM_017016610.1 Intron XP_016872099.1
XM_017016611.1 Intron XP_016872100.1
XM_017016612.1 Intron XP_016872101.1

View Full Product Details