Product Details

SNP ID
rs12566673
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:151403599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAATTGTAGAAAAAATTTTCTT[C/T]CCTTGAAGCTGGCAGTGAAAAATAA
Phenotype
MIM: 614787 MIM: 602177
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POGZ PubMed Links

Gene Details

Gene
POGZ
Gene Name
pogo transposable element with ZNF domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001194937.1 6310 UTR 3 NP_001181866.1
NM_001194938.1 6310 UTR 3 NP_001181867.1
NM_015100.3 6310 UTR 3 NP_055915.2
NM_145796.3 6310 UTR 3 NP_665739.3
NM_207171.2 6310 UTR 3 NP_997054.1
XM_005244999.2 6310 UTR 3 XP_005245056.1
XM_005245000.4 6310 UTR 3 XP_005245057.1
XM_005245001.2 6310 UTR 3 XP_005245058.1
XM_005245005.1 6310 UTR 3 XP_005245062.1
XM_005245006.4 6310 UTR 3 XP_005245063.1
XM_011509331.2 6310 UTR 3 XP_011507633.1
XM_017000744.1 6310 UTR 3 XP_016856233.1
XM_017000745.1 6310 UTR 3 XP_016856234.1
XM_017000746.1 6310 UTR 3 XP_016856235.1
XM_017000747.1 6310 UTR 3 XP_016856236.1
XM_017000748.1 6310 UTR 3 XP_016856237.1
XM_017000749.1 6310 UTR 3 XP_016856238.1
Gene
PSMB4
Gene Name
proteasome subunit beta 4
There are no transcripts associated with this gene.

View Full Product Details