Product Details
- SNP ID
-
rs3742189
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:110523072 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAGTTCAGTCTCAATACCTAAGTT[G/T]TTCCAAAAATCCTCTTTAATAATAC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RAB20
PubMed Links
Gene Details
- Gene
- RAB20
- Gene Name
- RAB20, member RAS oncogene family
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017817.2 |
1516 |
UTR 3 |
|
|
NP_060287.1 |
View Full Product Details