Product Details

SNP ID
rs34644105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2059464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGAGCCTACCACCAGGTCTCCC[A/T]CTCCCGCGGTGCCCTCCCTTCCCTT
Phenotype
MIM: 603825 MIM: 610016 MIM: 613487 MIM: 610963
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HIC1 PubMed Links

Gene Details

Gene
HIC1
Gene Name
hypermethylated in cancer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098202.1 2831 UTR 3 NP_001091672.1
NM_006497.3 2831 UTR 3 NP_006488.2
Gene
MIR132
Gene Name
microRNA 132
There are no transcripts associated with this gene.

Gene
MIR212
Gene Name
microRNA 212
There are no transcripts associated with this gene.

Gene
SMG6
Gene Name
SMG6, nonsense mediated mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256827.1 2831 Intron NP_001243756.1
NM_001256828.1 2831 Intron NP_001243757.1
NM_001282326.1 2831 Intron NP_001269255.1
NM_017575.4 2831 Intron NP_060045.4
XM_005256569.3 2831 Intron XP_005256626.1
XM_005256570.3 2831 Intron XP_005256627.1
XM_005256571.4 2831 Intron XP_005256628.1
XM_011523769.2 2831 Intron XP_011522071.1
XM_011523772.2 2831 Intron XP_011522074.1
XM_011523773.2 2831 Intron XP_011522075.1
XM_011523774.2 2831 Intron XP_011522076.1
XM_011523775.2 2831 Intron XP_011522077.1
XM_017024398.1 2831 Intron XP_016879887.1
XM_017024399.1 2831 Intron XP_016879888.1

View Full Product Details