Product Details

SNP ID
rs697636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51889089 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGACAGGCCTCATGGTCGCATGCT[A/T]CCACGGCTTCCAGAGTGTTGTGGCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD33 PubMed Links

Gene Details

Gene
ANKRD33
Gene Name
ankyrin repeat domain 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130015.1 517 Missense Mutation TAC,TTC Y5F NP_001123487.1
NM_001304459.1 517 Missense Mutation TAC,TTC Y5F NP_001291388.1
NM_001304460.1 517 Intron NP_001291389.1
NM_182608.3 517 Missense Mutation TAC,TTC Y140F NP_872414.3
XM_017019258.1 517 Missense Mutation TAC,TTC Y149F XP_016874747.1

View Full Product Details