Product Details
- SNP ID
-
rs11168827
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:49025894 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTTAGGATTAGGTTGGGTGAGGCA[C/G]ATGATAGTCTCAGAACTGTTTCCTT
- Phenotype
-
MIM: 602113
MIM: 602742
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KMT2D
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114640737] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KMT2D
- Gene Name
- lysine methyltransferase 2D
- Gene
- PRKAG1
- Gene Name
- protein kinase AMP-activated non-catalytic subunit gamma 1
There are no transcripts associated with this gene.
View Full Product Details