Product Details

SNP ID
rs10740420
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:74159932 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATATGACTCAATCTAGGAGTTCACC[A/G]CTTGTCTCTGTTGTCCTTTAGATTC
Phenotype
MIM: 102750 MIM: 610366
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADK PubMed Links

Gene Details

Gene
ADK
Gene Name
adenosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123.3 Intron NP_001114.2
NM_001202449.1 Intron NP_001189378.1
NM_001202450.1 Intron NP_001189379.1
NM_006721.3 Intron NP_006712.2
XM_017015698.1 Intron XP_016871187.1
XM_017015699.1 Intron XP_016871188.1
XM_017015700.1 Intron XP_016871189.1
XM_017015701.1 Intron XP_016871190.1
XM_017015702.1 Intron XP_016871191.1
XM_017015703.1 Intron XP_016871192.1
XM_017015704.1 Intron XP_016871193.1
XM_017015705.1 Intron XP_016871194.1
XM_017015706.1 Intron XP_016871195.1
Gene
AP3M1
Gene Name
adaptor related protein complex 3 mu 1 subunit
There are no transcripts associated with this gene.

View Full Product Details