Product Details
- SNP ID
-
rs17620012
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:63897759 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCTATTGCTATTTCACTACATTTTT[C/T]CTCCTTAGTTGGAAAAACCTTTTTA
- Phenotype
-
MIM: 191760
MIM: 614633
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
UGP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79983499] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UGP2
- Gene Name
- UDP-glucose pyrophosphorylase 2
There are no transcripts associated with this gene.
- Gene
- VPS54
- Gene Name
- VPS54, GARP complex subunit
View Full Product Details