Product Details

SNP ID
rs10746724
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:83749856 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCACCATACTGAATATAAACGTAG[A/G]AAGACTGCTGAGAAGCTCAGAAATG
Phenotype
MIM: 611356
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GKAP1 PubMed Links

Gene Details

Gene
GKAP1
Gene Name
G kinase anchoring protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135953.1 Intron NP_001129425.1
NM_025211.3 Intron NP_079487.2
XM_005252241.2 Intron XP_005252298.1
XM_006717293.1 Intron XP_006717356.1
XM_011519058.2 Intron XP_011517360.1
XM_017015171.1 Intron XP_016870660.1

View Full Product Details