Product Details

SNP ID
rs12528857
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46702199 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACATTTCAAAAACTATTTTGGTAC[A/C]TTTCAAATACAGTGTTTAAATTAAA
Phenotype
MIM: 601690 MIM: 611200
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
PLA2G7 PubMed Links

Gene Details

Gene
PLA2G7
Gene Name
phospholipase A2 group VII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168357.1 6767 Intron NP_001161829.1
NM_005084.3 6767 Intron NP_005075.3
XM_005249408.4 6767 Intron XP_005249465.1
Gene
TDRD6
Gene Name
tudor domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010870.2 6767 UTR 3 NP_001010870.1
NM_001168359.1 6767 UTR 3 NP_001161831.1

View Full Product Details