Product Details
- SNP ID
-
rs11885824
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:49920033 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TATTCAAGTAATGCAACCAATTTTT[A/C]CTGAAACATCAACTTGCAATGTAAT
- Phenotype
-
MIM: 600565
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NRXN1
PubMed Links
Gene Details
- Gene
- NRXN1
- Gene Name
- neurexin 1
View Full Product Details