Product Details

SNP ID
rs10755098
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:141884909 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AATGTCATATAACAACTGCAAAGAT[C/T]ATTACAGATTTTACTTCTTGGTTGA
Phenotype
MIM: 601867
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP1B3 PubMed Links

Gene Details

Gene
ATP1B3
Gene Name
ATPase Na+/K+ transporting subunit beta 3
There are no transcripts associated with this gene.

View Full Product Details