Product Details

SNP ID
rs3775488
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:73997135 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCAAGAGTAAGGAAATTCTCTAACA[C/T]ATTCCCCAAGAAACATTAAAATATT
Phenotype
MIM: 600324 MIM: 121010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXCL5 PubMed Links

Gene Details

Gene
CXCL5
Gene Name
C-X-C motif chemokine ligand 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002994.4 995 UTR 3 NP_002985.1
Gene
PPBP
Gene Name
pro-platelet basic protein
There are no transcripts associated with this gene.

View Full Product Details