Product Details
- SNP ID
-
rs3893143
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
53
- Location
-
Chr.1:236691872 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCATGGTGTGTTTGTGCACGTGTG[A/G]TAGGGAGATAATACCCGATGCTAAA
- Phenotype
-
MIM: 102573
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACTN2
PubMed Links
Gene Details
- Gene
- ACTN2
- Gene Name
- actinin alpha 2
View Full Product Details