Product Details

SNP ID
rs2279433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:45459359 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTAGGTAGGTAAGATTGGCTTTCC[A/G]CCCCACTTCTCCCTTCCCAAACTAG
Phenotype
MIM: 613335
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102724323 PubMed Links

Gene Details

Gene
LOC102724323
Gene Name
uncharacterized LOC102724323
There are no transcripts associated with this gene.

Gene
MARCH8
Gene Name
membrane associated ring-CH-type finger 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002266.2 Intron NP_001002266.1
NM_001282866.1 Intron NP_001269795.1
NM_145021.5 Intron NP_659458.2
XM_005271804.2 Intron XP_005271861.1
XM_006717704.2 Intron XP_006717767.1
XM_011539492.2 Intron XP_011537794.1
XM_011539493.2 Intron XP_011537795.1
XM_011539494.2 Intron XP_011537796.1
XM_011539495.1 Intron XP_011537797.1
XM_017015894.1 Intron XP_016871383.1

View Full Product Details