Product Details

SNP ID
rs7558470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:15005276 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAACTGTACACCTGTTGACTAGC[A/G]ACTCCCCTTACTCGCTCCCCAACAT
Phenotype
MIM: 608025
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NBAS PubMed Links

Gene Details

Gene
NBAS
Gene Name
neuroblastoma amplified sequence
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015909.3 Intron NP_056993.2
XM_011510357.2 Intron XP_011508659.1
XM_011510358.2 Intron XP_011508660.1
XM_011510359.2 Intron XP_011508661.1
XM_011510360.2 Intron XP_011508662.1
XM_011510361.2 Intron XP_011508663.1
XM_017004317.1 Intron XP_016859806.1

View Full Product Details