Product Details
- SNP ID
-
rs13243663
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:129152305 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTCATAGCAATACTATCCACAGCA[C/G]CCAAAAGGTGGGAGCAACACAAATG
- Phenotype
-
MIM: 610120
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TSPAN33
PubMed Links
Gene Details
- Gene
- TSPAN33
- Gene Name
- tetraspanin 33
View Full Product Details