Product Details
- SNP ID
-
rs9825088
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:190307811 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAGAAGAGTAAATGATATTTGAC[C/T]GTTACATATGTATATATAGACGAAA
- Phenotype
-
MIM: 603718
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN1
PubMed Links
Gene Details
- Gene
- CLDN1
- Gene Name
- claudin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021101.4 |
1342 |
UTR 3 |
|
|
NP_066924.1 |
View Full Product Details