Product Details

SNP ID
rs11604149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6498412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAACTCCTGCTAGCTGAGCTCC[A/G]AACTCTGTTCTCAGCTGTGTTGCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNHD1 PubMed Links

Gene Details

Gene
DNHD1
Gene Name
dynein heavy chain domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144666.2 761 Missense Mutation CAA,CGA Q66R NP_653267.2
NM_173589.3 761 Missense Mutation CAA,CGA Q66R NP_775860.3

View Full Product Details