Product Details

SNP ID
rs9884820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:118857581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGAAACAAGACAGAATTTTTTT[A/T]AAAAAAATCAAACCGTAAGTAATCT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SYNPO2 PubMed Links

Gene Details

Gene
SYNPO2
Gene Name
synaptopodin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128933.2 Intron NP_001122405.1
NM_001128934.2 Intron NP_001122406.1
NM_001286754.1 Intron NP_001273683.1
NM_001286755.1 Intron NP_001273684.1
NM_133477.2 Intron NP_597734.2

View Full Product Details