Product Details

SNP ID
rs1365772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:58602109 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCCTGTTCTTCATCAAACTTTCAA[C/T]GTATTCATTTATTTATTCCCTCATG
Phenotype
MIM: 602192
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM10 PubMed Links

Gene Details

Gene
ADAM10
Gene Name
ADAM metallopeptidase domain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110.3 Intron NP_001101.1
NM_001320570.1 Intron NP_001307499.1
XM_017021861.1 Intron XP_016877350.1

View Full Product Details