Product Details

SNP ID
rs814628
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:88669895 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CACAATCGACTTCATTGTAAAGAAA[A/G]CTGGACAGAAGCAGCTACACTATGT
Phenotype
MIM: 601980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIPF PubMed Links

Gene Details

Gene
LIPF
Gene Name
lipase F, gastric type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198828.1 537 Missense Mutation ACT,GCT T128A NP_001185757.1
NM_001198829.1 537 Missense Mutation ACT,GCT T171A NP_001185758.1
NM_001198830.1 537 Missense Mutation ACT,GCT T138A NP_001185759.1
NM_004190.3 537 Missense Mutation ACT,GCT T161A NP_004181.1
XM_011540311.1 537 Missense Mutation ACT,GCT T161A XP_011538613.1

View Full Product Details