Product Details

SNP ID
rs35335704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:105334254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAACTTCTAGGGCAACGCTGGGG[G/T]CAGCCAGCAATGGATGTTTGTAGAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C12orf75 PubMed Links

Gene Details

Gene
C12orf75
Gene Name
chromosome 12 open reading frame 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145199.1 Intron NP_001138671.1
Gene
KCCAT198
Gene Name
renal clear cell carcinoma-associated transcript 198
There are no transcripts associated with this gene.

View Full Product Details