Product Details

SNP ID
rs10035989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:159157983 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAGTTTCATATACATCAAAATA[C/T]TGAAGACTCCGTCATAAAGTTCAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF145 PubMed Links

Gene Details

Gene
RNF145
Gene Name
ring finger protein 145
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199380.1 2892 UTR 3 NP_001186309.1
NM_001199381.1 2892 UTR 3 NP_001186310.1
NM_001199382.1 2892 UTR 3 NP_001186311.1
NM_001199383.1 2892 UTR 3 NP_001186312.1
NM_144726.2 2892 UTR 3 NP_653327.1
XM_017009138.1 2892 UTR 3 XP_016864627.1

View Full Product Details