Product Details

SNP ID
rs34613921
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:157088993 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TGAGAATACCCTAGTAAGGGGGAAA[C/G]AAAAGCCAATAAAAATGCATTCATA
Phenotype
MIM: 606652
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HAVCR2 PubMed Links

Gene Details

Gene
HAVCR2
Gene Name
hepatitis A virus cellular receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032782.4 Intron NP_116171.3

View Full Product Details