Product Details

SNP ID
rs11959935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:127232634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATAACAAAAAGTAGTACAGGGT[A/C]AATGTCCCAAGAGAAGTACAAAACT
Phenotype
MIM: 612453
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC105379198 PubMed Links

Gene Details

Gene
LOC105379198
Gene Name
uncharacterized LOC105379198
There are no transcripts associated with this gene.

Gene
MEGF10
Gene Name
multiple EGF like domains 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256545.1 Intron NP_001243474.1
NM_001308119.1 Intron NP_001295048.1
NM_001308121.1 Intron NP_001295050.1
NM_032446.2 Intron NP_115822.1
XM_011543694.1 Intron XP_011541996.1
XM_017009987.1 Intron XP_016865476.1
XM_017009988.1 Intron XP_016865477.1

View Full Product Details