Product Details

SNP ID
rs3213113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:159315876 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATATTTAAATAGCATGAAGGCCCA[G/T]GGCAACTTGAGAGCTGGAAAATCTA
Phenotype
MIM: 161561
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL12B PubMed Links

Gene Details

Gene
IL12B
Gene Name
interleukin 12B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002187.2 1254 UTR 3 NP_002178.2

View Full Product Details