Product Details

SNP ID
rs1865015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:107861275 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGTGCACACGTGTGAATATGTGA[A/G]AGAGTGTAAGTGCCTGTACGTATAA
Phenotype
MIM: 609083
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXL17 PubMed Links

Gene Details

Gene
FBXL17
Gene Name
F-box and leucine rich repeat protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163315.2 2958 UTR 3 NP_001156787.2
XM_005272048.4 2958 Intron XP_005272105.1
XM_005272050.4 2958 Intron XP_005272107.1
XM_011543574.2 2958 Intron XP_011541876.1
XM_011543575.2 2958 Intron XP_011541877.1
XM_011543576.2 2958 Intron XP_011541878.1
XM_011543577.2 2958 Intron XP_011541879.1
XM_011543578.2 2958 Intron XP_011541880.1
XM_011543579.2 2958 Intron XP_011541881.1
XM_011543580.2 2958 Intron XP_011541882.1
XM_017009729.1 2958 Intron XP_016865218.1

View Full Product Details