Product Details

SNP ID
rs1053411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151663542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACACGAAGGGGAGGGTTAAAGA[C/G]AGAATCCGGTACTGTGGAAGGAGTG
Phenotype
MIM: 182120
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPARC PubMed Links

Gene Details

Gene
SPARC
Gene Name
secreted protein acidic and cysteine rich
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001309443.1 1144 UTR 3 NP_001296372.1
NM_001309444.1 1144 Silent Mutation CTC,CTG L313L NP_001296373.1
NM_003118.3 1144 UTR 3 NP_003109.1

View Full Product Details