Product Details

SNP ID
rs13387999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85584951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCCGCCTGGCACTCAGAACCCA[C/T]TGCTAAGAAGTGGTAGGAAATGTAA
Phenotype
MIM: 607029 MIM: 603177
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VAMP5 PubMed Links

Gene Details

Gene
VAMP5
Gene Name
vesicle associated membrane protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006634.2 Intron NP_006625.1
Gene
VAMP8
Gene Name
vesicle associated membrane protein 8
There are no transcripts associated with this gene.

View Full Product Details