Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_182526.2 | 3580 | UTR 3 | NP_872332.1 | ||
XM_005267373.2 | 3580 | UTR 3 | XP_005267430.1 | ||
XM_005267374.3 | 3580 | UTR 3 | XP_005267431.1 | ||
XM_005267375.2 | 3580 | UTR 3 | XP_005267432.1 | ||
XM_006720062.2 | 3580 | UTR 3 | XP_006720125.1 | ||
XM_017021051.1 | 3580 | UTR 3 | XP_016876540.1 | ||
XM_017021052.1 | 3580 | UTR 3 | XP_016876541.1 | ||
XM_017021053.1 | 3580 | UTR 3 | XP_016876542.1 | ||
XM_017021054.1 | 3580 | UTR 3 | XP_016876543.1 | ||
XM_017021055.1 | 3580 | UTR 3 | XP_016876544.1 |