Product Details

SNP ID
rs10180682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:120254457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTCCCTCTGCCCGAGGAATAG[A/G]ACATTTTCCCAGGATTGTGTGTGGA
Phenotype
MIM: 179551
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RALB PubMed Links

Gene Details

Gene
RALB
Gene Name
RALB Ras like proto-oncogene B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002881.2 Intron NP_002872.1
XM_005263724.1 Intron XP_005263781.1
XM_005263727.1 Intron XP_005263784.1
XM_005263728.1 Intron XP_005263785.1
XM_005263729.2 Intron XP_005263786.1
XM_011511573.1 Intron XP_011509875.1
XM_011511574.1 Intron XP_011509876.1
XM_017004621.1 Intron XP_016860110.1
XM_017004622.1 Intron XP_016860111.1

View Full Product Details