Product Details
- SNP ID
-
rs1437421
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:120218255 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTGGTACTTAACCTTAAGCTCATGG[A/C]AGACAGCTGAGGAAGTACTCGCAGC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM185B
PubMed Links
Gene Details
- Gene
- TMEM185B
- Gene Name
- transmembrane protein 185B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024121.2 |
5154 |
UTR 3 |
|
|
NP_077026.2 |
View Full Product Details