Product Details

SNP ID
rs11692738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:208375522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAACATTTCTTTTTATAGTTGAAC[C/T]AGAGGCAAGGCAAGGAACTTGACAT
Phenotype
MIM: 601469
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTH2R PubMed Links

Gene Details

Gene
PTH2R
Gene Name
parathyroid hormone 2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001309516.1 Intron NP_001296445.1
NM_005048.3 Intron NP_005039.1

View Full Product Details