Product Details

SNP ID
rs10176000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:105256525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTACAAGATAAAACAATGTAAA[C/T]GTCTAAAGAGATTCCATTTCTTTGA
Phenotype
MIM: 606237
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TGFBRAP1 PubMed Links

Gene Details

Gene
TGFBRAP1
Gene Name
transforming growth factor beta receptor associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142621.2 Intron NP_001136093.1
NM_004257.5 Intron NP_004248.2
XM_011512193.2 Intron XP_011510495.1
XM_017005339.1 Intron XP_016860828.1

View Full Product Details