Product Details
- SNP ID
-
rs6718426
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:112486675 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAGACAACGATTAAGATCTAGGGT[C/G]AAAAGTGTAAGTGTTCACCCCAGCT
- Phenotype
-
MIM: 608291
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TTL
PubMed Links
Gene Details
- Gene
- TTL
- Gene Name
- tubulin tyrosine ligase
View Full Product Details