Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001287499.1 | Intron | NP_001274428.1 | |||
NM_001287500.1 | Intron | NP_001274429.1 | |||
NM_001287501.1 | Intron | NP_001274430.1 | |||
NM_001287502.1 | Intron | NP_001274431.1 | |||
NM_152558.4 | Intron | NP_689771.3 | |||
XM_006715676.2 | Intron | XP_006715739.1 | |||
XM_011515242.1 | Intron | XP_011513544.1 | |||
XM_011515243.1 | Intron | XP_011513545.1 | |||
XM_011515244.2 | Intron | XP_011513546.1 | |||
XM_017011902.1 | Intron | XP_016867391.1 | |||
XM_017011903.1 | Intron | XP_016867392.1 |