Product Details

SNP ID
rs3092983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40739577 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTTTCACTAAAACTCAGGTGCTG[A/G]AACCAGACCCCAGGATATGAGTTAG
Phenotype
MIM: 179617 MIM: 611873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD51 PubMed Links

Gene Details

Gene
RAD51
Gene Name
RAD51 recombinase
There are no transcripts associated with this gene.

Gene
RMDN3
Gene Name
regulator of microtubule dynamics 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304802.1 Intron NP_001291731.1
NM_001323894.1 Intron NP_001310823.1
NM_001323895.1 Intron NP_001310824.1
NM_001323896.1 Intron NP_001310825.1
NM_001323897.1 Intron NP_001310826.1
NM_018145.2 Intron NP_060615.1
XM_011521755.2 Intron XP_011520057.1

View Full Product Details