Product Details

SNP ID
rs4980181
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:123147873 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TCCAAGGGGTCCAAGGAGTCGGAGT[C/T]GCCGCAGCCGCCAGGAACCCGGGCT
Phenotype
MIM: 603719 MIM: 600647
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BUB3 PubMed Links

Gene Details

Gene
BUB3
Gene Name
BUB3, mitotic checkpoint protein
There are no transcripts associated with this gene.

Gene
HMX2
Gene Name
H6 family homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005519.1 Intron NP_005510.1
XM_005269743.3 Intron XP_005269800.1

View Full Product Details