Product Details

SNP ID
rs13262300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:64374995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGAGATGATTGAAAAGGTTGTT[C/T]AGCTTTTGCCCCTGAATTTGTCATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724623 PubMed Links

Gene Details

Gene
LOC102724623
Gene Name
uncharacterized LOC102724623
There are no transcripts associated with this gene.

Gene
MIR124-2
Gene Name
microRNA 124-2
There are no transcripts associated with this gene.

Gene
MIR124-2HG
Gene Name
MIR124-2 host gene
There are no transcripts associated with this gene.

View Full Product Details