Product Details

SNP ID
rs4123220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:121642411 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCCGCCTCCTTTCTTTCTCTTGG[A/T]CCTTGCCAACAAGCCTGCTGATGAC
Phenotype
MIM: 601636 MIM: 614353
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HAS2 PubMed Links

Gene Details

Gene
HAS2
Gene Name
hyaluronan synthase 2
There are no transcripts associated with this gene.

Gene
HAS2-AS1
Gene Name
HAS2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details