Product Details

SNP ID
rs12997506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86027145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGGCCTCCTGCAGCACAGGTGA[A/G]GCCCCAGCCATCTCAGGGGGACTCA
Phenotype
MIM: 616404
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC90784 PubMed Links

Gene Details

Gene
LOC90784
Gene Name
uncharacterized LOC90784
There are no transcripts associated with this gene.

Gene
POLR1A
Gene Name
RNA polymerase I subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015425.4 5820 UTR 3 NP_056240.2

View Full Product Details